Product Details
- SNP ID
-
rs147502266
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57323751 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGTGGGGACAGTTGGATGCAGCC[C/T]AGAGAACCTTGTATCAGGAGGTGAT
- Phenotype
-
MIM: 616847
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF543
PubMed Links
Gene Details
- Gene
- ZNF543
- Gene Name
- zinc finger protein 543
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_213598.3 |
445 |
Nonsense Mutation |
CAG,TAG |
Q30* |
NP_998763.2 |
View Full Product Details