Product Details

SNP ID
rs147522098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48385603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAGGGAAGAAGACATCTGCCAG[C/T]CCCCTTTCAGTCCCCAGCTCCCTGA
Phenotype
MIM: 602717 MIM: 131235 MIM: 608373
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRIN2D PubMed Links

Gene Details

Gene
GRIN2D
Gene Name
glutamate ionotropic receptor NMDA type subunit 2D
There are no transcripts associated with this gene.

Gene
KDELR1
Gene Name
KDEL endoplasmic reticulum protein retention receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006801.2 Intron NP_006792.1
Gene
SYNGR4
Gene Name
synaptogyrin 4
There are no transcripts associated with this gene.

View Full Product Details