Product Details

SNP ID
rs147945859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58455524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAGGCAGCCCAGGACGCCTGAG[A/C]GGCAGAAGCCATGTGCACAGGAGGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF324B PubMed Links

Gene Details

Gene
ZNF324B
Gene Name
zinc finger protein 324B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207395.2 711 Silent Mutation AGG,CGG R194R NP_997278.2
XM_005258915.4 711 Silent Mutation AGG,CGG R194R XP_005258972.1
XM_005258918.4 711 Silent Mutation AGG,CGG R194R XP_005258975.1

View Full Product Details