Product Details

SNP ID
rs147988688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15110263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGAGGCCGCCAGGCTCCTGCG[C/T]GCCCTGGTGCTTGCGTGGGACCCTG
Phenotype
MIM: 605770
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ILVBL PubMed Links

Gene Details

Gene
ILVBL
Gene Name
ilvB acetolactate synthase like
There are no transcripts associated with this gene.

Gene
SYDE1
Gene Name
synapse defective Rho GTPase homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300910.1 1060 Silent Mutation CGC,CGT R263R NP_001287839.1
NM_033025.5 1060 Silent Mutation CGC,CGT R330R NP_149014.3

View Full Product Details