Product Details

SNP ID
rs148231062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53237101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTACCATGTATATTATGTTTACA[A/G]TGTTTCTTTTCAATGTGTATTTTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF677 PubMed Links

Gene Details

Gene
ZNF677
Gene Name
zinc finger protein 677
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317998.1 2038 Silent Mutation CAC,CAT H542H NP_001304927.1
NM_182609.3 2038 Silent Mutation CAC,CAT H542H NP_872415.1
XM_005258862.4 2038 Silent Mutation CAC,CAT H506H XP_005258919.1
XM_011526912.2 2038 Silent Mutation CAC,CAT H542H XP_011525214.1

View Full Product Details