Product Details

SNP ID
rs148782724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5720713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCCCCGTGGCGGTTGAGGGGCA[C/G]TGGTGGCGGCGGAAGCCCAAGTCGA
Phenotype
MIM: 605490
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CATSPERD PubMed Links

Gene Details

Gene
CATSPERD
Gene Name
cation channel sperm associated auxiliary subunit delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152784.3 329 UTR 5 NP_689997.3
XM_011527885.2 329 UTR 5 XP_011526187.1
XM_011527886.2 329 UTR 5 XP_011526188.1
XM_011527887.2 329 UTR 5 XP_011526189.1
XM_011527888.2 329 UTR 5 XP_011526190.1
XM_011527889.2 329 UTR 5 XP_011526191.1
XM_011527890.2 329 UTR 5 XP_011526192.1
XM_011527891.1 329 Intron XP_011526193.1
XM_011527892.1 329 Intron XP_011526194.1
XM_017026564.1 329 UTR 5 XP_016882053.1
XM_017026565.1 329 UTR 5 XP_016882054.1
XM_017026566.1 329 UTR 5 XP_016882055.1
Gene
LONP1
Gene Name
lon peptidase 1, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276479.1 329 Intron NP_001263408.1
NM_001276480.1 329 Intron NP_001263409.1
NM_004793.3 329 Intron NP_004784.2
XM_011528441.2 329 Intron XP_011526743.1

View Full Product Details