Product Details

SNP ID
rs149104673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38870662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGCCAGGTGATCCTGGAGGTCA[A/G]TAAGGAGGTGCCGCACATCCTGAAG
Phenotype
MIM: 604480
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RINL PubMed Links

Gene Details

Gene
RINL
Gene Name
Ras and Rab interactor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195833.1 1013 Missense Mutation ACT,ATT T311I NP_001182762.1
NM_198445.3 1013 Intron NP_940847.1
XM_006723015.2 1013 Missense Mutation ACT,ATT T311I XP_006723078.1
XM_006723016.2 1013 Missense Mutation ACT,ATT T197I XP_006723079.1
XM_011526454.2 1013 Missense Mutation ACT,ATT T311I XP_011524756.1
XM_011526455.2 1013 Intron XP_011524757.1
Gene
SIRT2
Gene Name
sirtuin 2
There are no transcripts associated with this gene.

View Full Product Details