Product Details

SNP ID
rs149174447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48741379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCGGCTTGCCAGCATTTTCTCG[A/G]GCTGCAGAGGCTGGAGGCTTATGGA
Phenotype
MIM: 211100 MIM: 609278 MIM: 609623
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FUT1 PubMed Links

Gene Details

Gene
FUT1
Gene Name
fucosyltransferase 1 (H blood group)
There are no transcripts associated with this gene.

Gene
IZUMO1
Gene Name
izumo sperm-egg fusion 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321864.1 964 Missense Mutation CCC,CTC P172L NP_001308793.1
NM_001321865.1 964 Missense Mutation CCC,CTC P111L NP_001308794.1
NM_182575.2 964 Missense Mutation CCC,CTC P285L NP_872381.2
XM_005258793.4 964 Missense Mutation CCC,CTC P311L XP_005258850.1
XM_011526811.2 964 Missense Mutation CCC,CTC P291L XP_011525113.1
XM_011526812.2 964 Missense Mutation CCC,CTC P285L XP_011525114.1
XM_011526813.2 964 Missense Mutation CCC,CTC P285L XP_011525115.1
XM_011526815.2 964 Missense Mutation CCC,CTC P172L XP_011525117.1
XM_017026646.1 964 Missense Mutation CCC,CTC P318L XP_016882135.1
Gene
RASIP1
Gene Name
Ras interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017805.2 964 Intron NP_060275.2
XM_011527053.2 964 Intron XP_011525355.2
XM_011527054.2 964 Intron XP_011525356.2
XM_011527055.2 964 Intron XP_011525357.2
XM_017026914.1 964 Intron XP_016882403.1
XM_017026915.1 964 Intron XP_016882404.1

View Full Product Details