Product Details

SNP ID
rs149336251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38932409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGATGCACCGCCTGGGGCCCCC[C/G]AAGCGGCCGCCTCGGAAGCTGGGCC
Phenotype
MIM: 609094 MIM: 603021 MIM: 612804
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBXO17 PubMed Links

Gene Details

Gene
FBXO17
Gene Name
F-box protein 17
There are no transcripts associated with this gene.

Gene
MRPS12
Gene Name
mitochondrial ribosomal protein S12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021107.1 436 Silent Mutation CCC,CCG P42P NP_066930.1
NM_033362.3 436 Silent Mutation CCC,CCG P42P NP_203526.1
NM_033363.1 436 Silent Mutation CCC,CCG P42P NP_203527.1
Gene
SARS2
Gene Name
seryl-tRNA synthetase 2, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details