Product Details

SNP ID
rs149391173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2427029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGCGTCCATGTAGCGGTCCAT[A/G]CACATGGCGATGCACTTCTGCGGGA
Phenotype
MIM: 150341 MIM: 607383 MIM: 610477
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMNB2 PubMed Links

Gene Details

Gene
LMNB2
Gene Name
lamin B2
There are no transcripts associated with this gene.

Gene
MIR7108
Gene Name
microRNA 7108
There are no transcripts associated with this gene.

Gene
TIMM13
Gene Name
translocase of inner mitochondrial membrane 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012458.3 590 Silent Mutation NP_036590.1
Gene
TMPRSS9
Gene Name
transmembrane protease, serine 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011527978.2 590 Intron XP_011526280.1
XM_011527980.1 590 Intron XP_011526282.1
XM_011527982.2 590 Intron XP_011526284.1

View Full Product Details