Product Details

SNP ID
rs149447725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43268125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCGTGACTTCGGCAGTGGTGGGC[G/T]GGTTCCAGAAGTTTAAAAGTGATGC
Phenotype
MIM: 176398
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PSG9 PubMed Links

Gene Details

Gene
PSG9
Gene Name
pregnancy specific beta-1-glycoprotein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301707.1 183 Missense Mutation NP_001288636.1
NM_001301708.1 183 Missense Mutation NP_001288637.1
NM_001301709.1 183 Missense Mutation NP_001288638.1
NM_002784.4 183 Missense Mutation NP_002775.3
XM_005259075.3 183 Missense Mutation XP_005259132.1
XM_017027004.1 183 Missense Mutation XP_016882493.1
XM_017027005.1 183 Missense Mutation XP_016882494.1
XM_017027006.1 183 Missense Mutation XP_016882495.1
XM_017027007.1 183 Missense Mutation XP_016882496.1

View Full Product Details