Product Details

SNP ID
rs149460582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54663032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGGGGCCCCTGGGAGGAGACG[C/T]CATGATCCCCACCTTCACGGCTCTG
Phenotype
MIM: 604821
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LILRB4 PubMed Links

Gene Details

Gene
LILRB4
Gene Name
leukocyte immunoglobulin like receptor B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278426.3 238 UTR 5 NP_001265355.2
NM_001278427.3 238 UTR 5 NP_001265356.2
NM_001278428.3 238 UTR 5 NP_001265357.2
NM_001278429.3 238 Missense Mutation GCC,GTC A41V NP_001265358.2
NM_001278430.3 238 Intron NP_001265359.2
XM_017026215.1 238 Missense Mutation GCC,GTC A41V XP_016881704.1
XM_017026216.1 238 Missense Mutation GCC,GTC A41V XP_016881705.1
XM_017026217.1 238 Missense Mutation GCC,GTC A41V XP_016881706.1

View Full Product Details