Product Details

SNP ID
rs149531003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2851484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTTCAGATGATGAAACTCAATT[C/T]AAGGCCAGTGGGTCAGTTTCTCAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF555 PubMed Links

Gene Details

Gene
ZNF555
Gene Name
zinc finger protein 555
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172775.1 288 Silent Mutation TTC,TTT F49F NP_001166246.1
NM_152791.4 288 Silent Mutation TTC,TTT F49F NP_690004.4
XM_011527716.2 288 Silent Mutation TTC,TTT F51F XP_011526018.1
XM_017026375.1 288 Silent Mutation TTC,TTT F51F XP_016881864.1

View Full Product Details