Product Details

SNP ID
rs149627321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17876023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGCTAAGTGGCTTCTGGGTTGT[A/C]CTGGCACGCGGTGTCATGCTTGTGG
Phenotype
MIM: 601843
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC5A5 PubMed Links

Gene Details

Gene
SLC5A5
Gene Name
solute carrier family 5 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000453.2 1541 Silent Mutation GTA,GTC V205V NP_000444.1
XM_011528192.2 1541 Silent Mutation GTA,GTC V205V XP_011526494.1
XM_011528193.2 1541 Silent Mutation GTA,GTC V116V XP_011526495.1
XM_011528194.2 1541 Silent Mutation GTA,GTC V83V XP_011526496.1
XM_017027158.1 1541 Silent Mutation GTA,GTC V116V XP_016882647.1

View Full Product Details