Product Details
- SNP ID
-
rs149630391
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57323808 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACCTGCGGACTTCTCATGTCTCTG[A/G]GTAAGGCCCCTTCTGGTCCTGTGCT
- Phenotype
-
MIM: 616847
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF543
PubMed Links
Gene Details
- Gene
- ZNF543
- Gene Name
- zinc finger protein 543
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_213598.3 |
502 |
Missense Mutation |
AGC,GGC |
S49G |
NP_998763.2 |
View Full Product Details