Product Details
- SNP ID
-
rs149689841
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:19120062 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGAGTAAGCAGGCCACCCAGAGC[C/G]CCGGCAATCCGCGCTGCAGTCTGCT
- Phenotype
-
MIM: 610823
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC25A42
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs756791] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC25A42
- Gene Name
- solute carrier family 25 member 42
There are no transcripts associated with this gene.
- Gene
- TMEM161A
- Gene Name
- transmembrane protein 161A
View Full Product Details