Product Details

SNP ID
rs150096976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38932350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCAGGCCCAGCTCTGGTTCCC[C/T]GGCTCTGGGCTACCTGCTCCATGGC
Phenotype
MIM: 609094 MIM: 603021 MIM: 612804
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO17 PubMed Links

Gene Details

Gene
FBXO17
Gene Name
F-box protein 17
There are no transcripts associated with this gene.

Gene
MRPS12
Gene Name
mitochondrial ribosomal protein S12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021107.1 377 Missense Mutation CGG,TGG R23W NP_066930.1
NM_033362.3 377 Missense Mutation CGG,TGG R23W NP_203526.1
NM_033363.1 377 Missense Mutation CGG,TGG R23W NP_203527.1
Gene
SARS2
Gene Name
seryl-tRNA synthetase 2, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details