Product Details

SNP ID
rs150123169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2389806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCATGGAGCCCACTGTGGCTGA[C/T]GTACACCTCGTGCCCAGGACAACCA
Phenotype
MIM: 610477
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS9 PubMed Links

Gene Details

Gene
TMPRSS9
Gene Name
transmembrane protease, serine 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011527978.2 282 Silent Mutation GAC,GAT D7D XP_011526280.1
XM_011527980.1 282 Intron XP_011526282.1
XM_011527982.2 282 Intron XP_011526284.1

View Full Product Details