Product Details
- SNP ID
-
rs150407533
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57159202 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACACCAGGTTGATCTTGCCCCTGG[A/C]TCTGGCTTGATTCTAAAGTCTCAGC
- Phenotype
-
MIM: 611168
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DUXA
PubMed Links
Gene Details
- Gene
- DUXA
- Gene Name
- double homeobox A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001012729.1 |
302 |
Missense Mutation |
AGC,ATC |
S86I |
NP_001012747.1 |
View Full Product Details