Product Details

SNP ID
rs150537373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6678175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGGGCTTCTCTCCCCAGAAATC[A/G]GAGGAGAGACCCCACATGAGGTAGT
Phenotype
MIM: 120700 MIM: 604520
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3 PubMed Links

Gene Details

Gene
C3
Gene Name
complement component 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000064.3 4920 Silent Mutation TCC,TCT S1609S NP_000055.2
Gene
TNFSF14
Gene Name
tumor necrosis factor superfamily member 14
There are no transcripts associated with this gene.

View Full Product Details