Product Details

SNP ID
rs151052094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16553882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTGACGGGGAGGAGGTGCTTCC[C/T]GGCTTGCTGGTTTGCATCGTACTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC35E1 PubMed Links

Gene Details

Gene
SLC35E1
Gene Name
solute carrier family 35 member E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024881.4 1048 Missense Mutation AGG,GGG R344G NP_079157.3

View Full Product Details