Product Details

SNP ID
rs151206783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57572628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGTGAATTAACTGGTGCTGAA[G/T]GAGGCCACACTTTAGGCTAAATGAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF416 PubMed Links

Gene Details

Gene
ZNF416
Gene Name
zinc finger protein 416
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017879.1 1447 Missense Mutation ATT,CTT I426L NP_060349.1
XM_005259051.3 1447 Missense Mutation ATT,CTT I354L XP_005259108.1

View Full Product Details