Product Details

SNP ID
rs111372554
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:167054441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAAAGTTCTCTTAGCTGCTCTG[A/G]TGGCTCCTCATAGGCTTCCCGGTTC
Phenotype
MIM: 602171
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPA33 PubMed Links

Gene Details

Gene
GPA33
Gene Name
glycoprotein A33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005814.2 1025 Missense Mutation CCA,TCA P285S NP_005805.1
XM_017000005.1 1025 Missense Mutation CCA,TCA P177S XP_016855494.1

View Full Product Details