Product Details

SNP ID
rs111493759
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:155320805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATATATCAAACCCTCTAGGTGG[A/G]TGACAAAAGACGCTAAACGCTGGGG
Phenotype
MIM: 134629
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FDPS PubMed Links

Gene Details

Gene
FDPS
Gene Name
farnesyl diphosphate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135821.1 961 Intron NP_001129293.1
NM_001135822.1 961 Intron NP_001129294.1
NM_001242824.1 961 Intron NP_001229753.1
NM_001242825.1 961 Intron NP_001229754.1
NM_002004.3 961 Intron NP_001995.1
XM_005244962.1 961 Intron XP_005245019.1
XM_005244963.1 961 Intron XP_005245020.1
Gene
LOC105371451
Gene Name
uncharacterized LOC105371451
There are no transcripts associated with this gene.

Gene
RUSC1
Gene Name
RUN and SH3 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105203.1 961 Intron NP_001098673.1
NM_001105204.1 961 Intron NP_001098674.1
NM_001105205.1 961 Intron NP_001098675.1
NM_001278227.1 961 Intron NP_001265156.1
NM_001278228.1 961 Intron NP_001265157.1
NM_001278229.1 961 Intron NP_001265158.1
NM_001278230.1 961 Intron NP_001265159.1
NM_014328.4 961 Intron NP_055143.2
XM_006711254.1 961 Intron XP_006711317.1
XM_006711256.1 961 Intron XP_006711319.1
XM_006711257.1 961 Intron XP_006711320.1
XM_017000891.1 961 Intron XP_016856380.1
XM_017000892.1 961 Intron XP_016856381.1
Gene
RUSC1-AS1
Gene Name
RUSC1 antisense RNA 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039517.1 961 Silent Mutation CAC,CAT H228H NP_001034606.1

View Full Product Details