Product Details

SNP ID
rs111575164
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:11108196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCACTCACCAGCCAATATAGCAC[C/T]GGCAGAGGTTTTCATGGGATGTCGC
Phenotype
MIM: 605960 MIM: 601231
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOSC10 PubMed Links

Gene Details

Gene
EXOSC10
Gene Name
exosome component 10
There are no transcripts associated with this gene.

Gene
LOC105376736
Gene Name
uncharacterized LOC105376736
There are no transcripts associated with this gene.

Gene
MTOR
Gene Name
mechanistic target of rapamycin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004958.3 7203 Missense Mutation CAG,CGG Q2540R NP_004949.1
XM_005263438.2 7203 Missense Mutation CAG,CGG Q2540R XP_005263495.1
XM_011541166.2 7203 Intron XP_011539468.1
XM_017000900.1 7203 Missense Mutation CAG,CGG Q2313R XP_016856389.1
XM_017000901.1 7203 Missense Mutation CAG,CGG Q2124R XP_016856390.1
XM_017000902.1 7203 Intron XP_016856391.1

View Full Product Details