Product Details
- SNP ID
-
rs111899359
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:183023738 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGGATGAGAGGGAGCCATCGGGCC[A/G]CGCCGGCCCTGCGGCCCCGGGGGCG
- Phenotype
-
MIM: 150290
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LAMC1
PubMed Links
Gene Details
- Gene
- LAMC1
- Gene Name
- laminin subunit gamma 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002293.3 |
279 |
Missense Mutation |
ACG,GCG |
T8A |
NP_002284.3 |
View Full Product Details