Product Details

SNP ID
rs111977166
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:28579770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCCAACCATCTCTGAAACCTGT[C/T]TGAAACTAGCCCACATGCCATATAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNHG12 PubMed Links

Gene Details

Gene
SNHG12
Gene Name
small nucleolar RNA host gene 12
There are no transcripts associated with this gene.

Gene
SNORA16A
Gene Name
small nucleolar RNA, H/ACA box 16A
There are no transcripts associated with this gene.

Gene
SNORA44
Gene Name
small nucleolar RNA, H/ACA box 44
There are no transcripts associated with this gene.

Gene
SNORA61
Gene Name
small nucleolar RNA, H/ACA box 61
There are no transcripts associated with this gene.

Gene
SNORD99
Gene Name
small nucleolar RNA, C/D box 99
There are no transcripts associated with this gene.

Gene
TRNAU1AP
Gene Name
tRNA selenocysteine 1 associated protein 1
There are no transcripts associated with this gene.

View Full Product Details