Product Details

SNP ID
rs112217438
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:93993153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCATTTACGCTGGCCAGTCCAT[G/T]TGGATGACCATATGGGCACAGGCTC
Phenotype
MIM: 601691
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ABCA4 PubMed Links

Gene Details

Gene
ABCA4
Gene Name
ATP binding cassette subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000350.2 7010 UTR 3 NP_000341.2

View Full Product Details