Product Details

SNP ID
rs112452625
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:22120197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGCACAGCAGCTCACAGCCGTC[A/G]ATGGCCTTGGACGTCTTGTTGCATG
Phenotype
MIM: 603490
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WNT4 PubMed Links

Gene Details

Gene
WNT4
Gene Name
Wnt family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030761.4 1082 Silent Mutation ATC,ATT I303I NP_110388.2
XM_011541597.2 1082 Silent Mutation ATC,ATT I325I XP_011539899.1
XM_011541598.2 1082 Silent Mutation ATC,ATT I248I XP_011539900.1
XM_011541599.1 1082 Intron XP_011539901.1

View Full Product Details