Product Details

SNP ID
rs112957416
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:32275353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGTGGAAGGCGCAGTCCCTGA[C/T]CACGGGCCAGGAAGGCTTCATCCCC
Phenotype
MIM: 153390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCK PubMed Links

Gene Details

Gene
LCK
Gene Name
LCK proto-oncogene, Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042771.2 604 Missense Mutation ACC,ATC T104I NP_001036236.1
NM_005356.4 604 Missense Mutation ACC,ATC T104I NP_005347.3
XM_011541453.2 604 Missense Mutation ACC,ATC T162I XP_011539755.1
XM_011541454.2 604 Missense Mutation ACC,ATC T162I XP_011539756.1
XM_011541455.2 604 Missense Mutation ACC,ATC T104I XP_011539757.1

View Full Product Details