Product Details

SNP ID
rs113681668
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:1218478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACCTCCAGCTCCTCGGCGGTCA[C/T]GATGCCGTCGTGGTTGGAGTCAATG
Phenotype
MIM: 614282 MIM: 600315
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SDF4 PubMed Links

Gene Details

Gene
SDF4
Gene Name
stromal cell derived factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016176.3 1221 Missense Mutation NP_057260.2
NM_016547.2 1221 Missense Mutation NP_057631.1
XM_011541556.1 1221 Intron XP_011539858.1
Gene
TNFRSF4
Gene Name
TNF receptor superfamily member 4
There are no transcripts associated with this gene.

View Full Product Details