Product Details

SNP ID
rs114096998
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:97078987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCACAAATCACCTTAACACACCG[G/T]ATTCACAGATAAGGGTACGCCTCTC
Phenotype
MIM: 612779
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DPYD PubMed Links

Gene Details

Gene
DPYD
Gene Name
dihydropyrimidine dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000110.3 3414 Missense Mutation GCG,TCG A1023S NP_000101.2
NM_001160301.1 3414 Intron NP_001153773.1
XM_005270562.3 3414 Missense Mutation GCG,TCG A951S XP_005270619.2
XM_006710397.3 3414 Intron XP_006710460.1
XM_017000507.1 3414 Missense Mutation GCG,TCG A986S XP_016855996.1
XM_017000508.1 3414 Missense Mutation GCG,TCG A858S XP_016855997.1
XM_017000509.1 3414 Missense Mutation GCG,TCG A858S XP_016855998.1
XM_017000510.1 3414 Missense Mutation GCG,TCG A858S XP_016855999.1

View Full Product Details