Product Details

SNP ID
rs114135727
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:248681394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACTTTTGCAGAAAATATATCTTC[A/T]CAAAGAGTCTCCACATGGCTGTTTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR14I1 PubMed Links

Gene Details

Gene
OR14I1
Gene Name
olfactory receptor family 14 subfamily I member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004734.1 1046 Missense Mutation GAG,GTG E304V NP_001004734.1
XM_005273132.3 1046 Missense Mutation GAG,GTG E304V XP_005273189.1

View Full Product Details