Product Details

SNP ID
rs114371433
Assay Type
Functionally Tested
NCBI dbSNP Submissions
11
Location
Chr.1:3865292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTGCAGGTGTGAACCAGCCTTG[C/T]GTTCCCACTTGGCCGTGGTGATATT
Phenotype
MIM: 616690 MIM: 601883
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CEP104 PubMed Links

Gene Details

Gene
CEP104
Gene Name
centrosomal protein 104
There are no transcripts associated with this gene.

Gene
DFFB
Gene Name
DNA fragmentation factor subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282669.1 Intron NP_001269598.1
NM_001320132.1 Intron NP_001307061.1
NM_001320136.1 Intron NP_001307065.1
NM_004402.3 Intron NP_004393.1
XM_011540865.2 Intron XP_011539167.1
XM_017000498.1 Intron XP_016855987.1
XM_017000499.1 Intron XP_016855988.1
XM_017000500.1 Intron XP_016855989.1

View Full Product Details