Product Details

SNP ID
rs114613721
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:193127494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAGCACCAGCTCCGTCTTTAAGG[G/T]TATTTTAACTCAGATCTTTAAAAAC
Phenotype
MIM: 607393
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDC73 PubMed Links

Gene Details

Gene
CDC73
Gene Name
cell division cycle 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024529.4 Intron NP_078805.3
XM_006711537.3 Intron XP_006711600.1
Gene
MIR1278
Gene Name
microRNA 1278
There are no transcripts associated with this gene.

View Full Product Details