Product Details

SNP ID
rs114842251
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:12775001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCACTGTTCATGGAGGCCTTTA[C/T]CAGGAGATGCTGCGAGACCCTGACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRAMEF12 PubMed Links

Gene Details

Gene
PRAMEF12
Gene Name
PRAME family member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080830.4 154 Missense Mutation ACC,ATC T45I NP_001074299.2

View Full Product Details