Product Details

SNP ID
rs114900375
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:92238738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCCTGCAGTTATTCCTAACATT[A/G]TACCCAAATTAGCTGACTTGTGTAT
Phenotype
MIM: 601749
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf146 PubMed Links

Gene Details

Gene
C1orf146
Gene Name
chromosome 1 open reading frame 146
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012425.1 Intron NP_001012425.1
XM_011541447.2 Intron XP_011539749.1
Gene
GLMN
Gene Name
glomulin, FKBP associated protein
There are no transcripts associated with this gene.

View Full Product Details