Product Details

SNP ID
rs115041629
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:182599082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCAGGAGTTGCCATCAGGAAAT[A/G]TTTGTTCAGTGATGCCCTGAGGTGA
Phenotype
MIM: 602514 MIM: 180435
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RGS16 PubMed Links

Gene Details

Gene
RGS16
Gene Name
regulator of G-protein signaling 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002928.3 1973 UTR 3 NP_002919.3
Gene
RNASEL
Gene Name
ribonuclease L
There are no transcripts associated with this gene.

View Full Product Details