Product Details

SNP ID
rs115220254
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:23780333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATTTCATACAGTAATGATCTTA[C/T]GTGTTTTGTTGACATGGAATTGCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PITHD1 PubMed Links

Gene Details

Gene
PITHD1
Gene Name
PITH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020362.4 Intron NP_065095.2
XM_011541804.1 Intron XP_011540106.1
Gene
TCEB3-AS1
Gene Name
TCEB3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details