Product Details

SNP ID
rs115324519
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:201955589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGTCTTTGCATTTCTCTTGCCC[C/T]CCTGCCCACTGCCCTCCTTCTCCCT
Phenotype
MIM: 602715 MIM: 605057
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LMOD1 PubMed Links

Gene Details

Gene
LMOD1
Gene Name
leiomodin 1
There are no transcripts associated with this gene.

Gene
TIMM17A
Gene Name
translocase of inner mitochondrial membrane 17 homolog A (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006335.2 Intron NP_006326.1

View Full Product Details