Product Details

SNP ID
rs115540438
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:222558798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAAAAGTTAAGCAAAATAAAAAG[G/T]TTTAATACTCATCACATTTTTCAAG
Phenotype
MIM: 604903
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TAF1A PubMed Links

Gene Details

Gene
TAF1A
Gene Name
TATA-box binding protein associated factor, RNA polymerase I subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201536.1 Intron NP_001188465.1
NM_005681.3 Intron NP_005672.1
NM_139352.2 Intron NP_647603.1
XM_005273343.3 Intron XP_005273400.1
XM_006711612.1 Intron XP_006711675.1
XM_006711613.3 Intron XP_006711676.1
XM_017002758.1 Intron XP_016858247.1
XM_017002759.1 Intron XP_016858248.1
XM_017002760.1 Intron XP_016858249.1

View Full Product Details