Product Details

SNP ID
rs115569131
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:233616642 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAAGTTAGCACATCAATGAATTC[C/T]TGTTATATGAATACAACTATTCTAA
Phenotype
MIM: 601745
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNK1 PubMed Links

Gene Details

Gene
KCNK1
Gene Name
potassium two pore domain channel subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002245.3 Intron NP_002236.1
XM_011544184.2 Intron XP_011542486.1
Gene
MIR4427
Gene Name
microRNA 4427
There are no transcripts associated with this gene.

View Full Product Details