Product Details

SNP ID
rs115695145
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:22712825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAAATGACTCATGCGTTGGCCC[A/G]CTATACATTGGGATGTTTACAGTAT
Phenotype
MIM: 600997
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPHB2 PubMed Links

Gene Details

Gene
EPHB2
Gene Name
EPH receptor B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001309192.1 Intron NP_001296121.1
NM_001309193.1 Intron NP_001296122.1
NM_004442.7 Intron NP_004433.2
NM_017449.4 Intron NP_059145.2
XM_006710441.3 Intron XP_006710504.1
XM_006710442.3 Intron XP_006710505.1
Gene
MIR4684
Gene Name
microRNA 4684
There are no transcripts associated with this gene.

View Full Product Details