Product Details

SNP ID
rs115786820
Assay Type
Functionally Tested
NCBI dbSNP Submissions
18
Location
Chr.1:158562815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTGTGGTTGAAGGTGTACATGG[A/C]CCGGGGCCGTGCATAGGTGAAGAGA
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
OR6P1 PubMed Links

Gene Details

Gene
OR6P1
Gene Name
olfactory receptor family 6 subfamily P member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160325.1 790 Missense Mutation GCC,TCC A264S NP_001153797.1

View Full Product Details