Product Details
- SNP ID
-
rs115971123
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:152154100 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAGGGAGAAGTGTTCTACCTTTCA[A/G]TGATTCAACCAGGTCTCATCTACAC
- Phenotype
-
MIM: 613259
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RPTN
PubMed Links
Gene Details
- Gene
- RPTN
- Gene Name
- repetin
View Full Product Details