Product Details

SNP ID
rs116084080
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:156466127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCTTCGGCTGCCCCTGGGCCCC[C/T]TGCCCACTGCCCCACTGAGGAGGCA
Phenotype
MIM: 600663
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf61 PubMed Links

Gene Details

Gene
C1orf61
Gene Name
chromosome 1 open reading frame 61
There are no transcripts associated with this gene.

Gene
MEF2D
Gene Name
myocyte enhancer factor 2D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271629.1 3551 UTR 3 NP_001258558.1
NM_005920.3 3551 UTR 3 NP_005911.1
XM_005245169.4 3551 UTR 3 XP_005245226.1
XM_005245170.3 3551 UTR 3 XP_005245227.1
XM_006711330.3 3551 UTR 3 XP_006711393.1
XM_006711332.3 3551 UTR 3 XP_006711395.1
XM_006711333.2 3551 UTR 3 XP_006711396.1
XM_006711334.3 3551 UTR 3 XP_006711397.1
XM_011509569.2 3551 UTR 3 XP_011507871.1
XM_017001314.1 3551 UTR 3 XP_016856803.1
XM_017001315.1 3551 UTR 3 XP_016856804.1

View Full Product Details