Product Details

SNP ID
rs116667331
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:155023909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGCTGCCCACAGCCCAGGCAG[G/T]AGGCCTGATGCAGCCAAAAGTGGCT
Phenotype
MIM: 607646
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DCST1 PubMed Links

Gene Details

Gene
DCST1
Gene Name
DC-STAMP domain containing 1
There are no transcripts associated with this gene.

Gene
DCST2
Gene Name
DC-STAMP domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144622.2 1336 Missense Mutation TAC,TCC Y598S NP_653223.2
XM_011509188.2 1336 Missense Mutation TAC,TCC Y378S XP_011507490.1
XM_011509189.2 1336 Missense Mutation TAC,TCC Y266S XP_011507491.1
Gene
ZBTB7B
Gene Name
zinc finger and BTB domain containing 7B
There are no transcripts associated with this gene.

View Full Product Details