Product Details

SNP ID
rs117761881
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:113764145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAAAACACCCTCAAAGGCAGGT[A/C]CCGTTAAAAACTAGTCTGGATAAAC
Phenotype
MIM: 604950 MIM: 615858
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PHTF1 PubMed Links

Gene Details

Gene
PHTF1
Gene Name
putative homeodomain transcription factor 1
There are no transcripts associated with this gene.

Gene
RSBN1
Gene Name
round spermatid basic protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018364.4 4308 UTR 3 NP_060834.2
XM_017001518.1 4308 Intron XP_016857007.1

View Full Product Details