Product Details

SNP ID
rs137984871
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:97078193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGACTATTAAAACCAGTCAGAG[A/C]CCGTATGTGCACAGCAAAAGAGTGG
Phenotype
MIM: 612779
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DPYD PubMed Links

Gene Details

Gene
DPYD
Gene Name
dihydropyrimidine dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000110.3 4208 UTR 3 NP_000101.2
NM_001160301.1 4208 Intron NP_001153773.1
XM_005270562.3 4208 UTR 3 XP_005270619.2
XM_006710397.3 4208 Intron XP_006710460.1
XM_017000507.1 4208 UTR 3 XP_016855996.1
XM_017000508.1 4208 UTR 3 XP_016855997.1
XM_017000509.1 4208 UTR 3 XP_016855998.1
XM_017000510.1 4208 UTR 3 XP_016855999.1

View Full Product Details