Product Details

SNP ID
rs138204394
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:247712055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCACGTGGAGAAGGCTTTGCTC[C/T]GGCCACTGGCAGAGGGGATCCTGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6F1 PubMed Links

Gene Details

Gene
OR6F1
Gene Name
olfactory receptor family 6 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005286.1 701 Missense Mutation CAG,CGG Q234R NP_001005286.1

View Full Product Details